Dishes, aluminium, disposable
Lieferant: Hach
2164000
HACH2164000EA 87.9 EUR
HACH2164000
Dishes, aluminium, disposable
Schiffchen Wägeschiffchen
Made of aluminium.
  • Crimped sides for stability
  • Include tabs for easy handling
  • Sturdy, with a flat bottom and slight taper for easy stacking
  • Temperature resistant up to +426 °C
Dishes are ideal for evaporating and general utility use.
Melting point: +676 °C.