Methylviolett 1 g/l in wässriger Lösung Indikator
Lieferant: Hach
|
Achtung
|
Synonyme:
Fleisher's Indicator Solution
2193432
HACH2193432EA
23.7
EUR
HACH2193432
Methylviolett 1 g/l in wässriger Lösung Indikator
Methylviolett
Formel:
C₆₇H₆₂N₈Na₄O₁₄S₃ Molecular Weight: 1391,41 g/mol |
MDL:
MFCD00070843 CAS-Nummer: 1340-02-9 EINECS: 207-776-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...