Nitrite test kit, colour disc, NI-6
Lieferant: Hach
224000
HACH224000EA
262
EUR
HACH224000
Nitrite test kit, colour disc, NI-6
Umwelt-Prüfkits
The kit uses the colour disc or ferrous sulphate method to measure nitrogen as nitrite NO²⁻.
- Simple, cost effective
- Portable test kit for analysis in the field
Lieferumfang: Kit includes colour disc, comparator box and tubes, reagents, deionised dilution water, instructions and carrying case.