Calcium Standard Solution, 1000 mg/L as Ca (NIST), 500 ml, Hach
Lieferant: Hach
2240349
HACH2240349EA 57.2 EUR
HACH2240349
Calcium Standard Solution, 1000 mg/L as Ca (NIST), 500 ml, Hach
Standards Kationischer Standards Calcium-Standards
For accuracy checks and instrument calibrations in calcium determinations.