Visual colour kits, phosphate
Lieferant: Hach
225001
HACH225001EA 429 EUR
HACH225001
Visual colour kits, phosphate
Umwelt-Prüfkits
This kit provides measurement of ortho- (reactive) and meta- (condensed) phosphate (requires hot acid digestion).
Lieferumfang: Kit includes five reagents, colour disc, comparator box, viewing adapter, two viewing tubes, bottle, flask, dropper tube, stove and cover, test tube clamp, funnel, filter paper, deionized water, instructions and carrying case.