Ferrozine mono-sodium salt hydrate

Lieferant: Hach

Synonyme: 5,6-Diphenyl-3-(2-pyridyl)-1,2,4-triazin-4,4''-disulfonsäure Mononatriumsalz Hydrat

230424
HACH230424EA 187 EUR
HACH230424
Ferrozine mono-sodium salt hydrate
Ferrozine mono-sodium salt hydrate
Formel: C₂₀H₁₃N₄NaO₆S₂·H₂O
Molecular Weight: 492,47 g/mol (anhy)
MDL: MFCD00150794
CAS-Nummer: 63451-29-6
EINECS: 274-196-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR