Buffer, alkali solution (3,0 N)
Lieferant: Hach
2307242
HACH2307242EA 24.4 EUR
HACH2307242
Buffer, alkali solution (3,0 N)
Puffer Assay-Puffer
These buffers are used in the determination of calcium and magnesium in brine.
Verpackung: Buffers are packed in a 100 ml bottle.