Ascorbic acid titrant -Lösung
Lieferant: Hach
Achtung
2308232
HACH2308232EA 28.7 EUR
HACH2308232
Ascorbic acid titrant -Lösung
Ascorbic acid titrant
This titrant is used for the determination of ascorbic acid in juice based beverages, by drop-count titration.