Bromocresol green-methyl red -Lösung pH-Indikator
Lieferant: Hach
Synonyme: Methyl Red - Bromocresol green solution
2329249 2329232
HACH2329249EA 38.4 EUR
HACH2329249 HACH2329232
Bromocresol green-methyl red -Lösung pH-Indikator
Bromocresol green-methyl red
Bromcresol green-methyl red pH indicator solutions are used to determine alkalinity of water by titration method. APHA formulation.