Bromocresol green-methyl red -Lösung pH-Indikator
Lieferant: Hach
Synonyme:
Methyl Red - Bromocresol green solution
2329249
2329232
HACH2329249EA
38.4
EUR
HACH2329249
HACH2329232
Bromocresol green-methyl red -Lösung pH-Indikator
Bromocresol green-methyl red
Bromcresol green-methyl red pH indicator solutions are used to determine alkalinity of water by titration method. APHA formulation.