Dissolved oxygen, reagent set
Lieferant: Hach
2351400
HACH2351400EA 226 EUR
HACH2351400
Dissolved oxygen, reagent set
Umwelt-Prüfkits
For determination of dissolved oxygen by the Winkler method (azide modification) and buret titration.