Silica reagents for 5000 silica analyzer
Lieferant: Hach
199503 2347003 2353103 2100803 4562700
HACH199503EA 139 EUR
HACH199503 HACH2347003 HACH2353103 HACH2100803 HACH4562700
Silica reagents for 5000 silica analyzer
Umwelt-Prüfkits
One unit each of the reagents is sufficient for 30 days of operation. The standard solution is sufficient for ten calibrations.