Silica reagents for 5000 silica analyzer
Lieferant: Hach
199503
2347003
2353103
2100803
4562700
HACH199503EA
139
EUR
HACH199503
HACH2347003
HACH2353103
HACH2100803
HACH4562700
Silica reagents for 5000 silica analyzer
Umwelt-Prüfkits
One unit each of the reagents is sufficient for 30 days of operation. The standard solution is sufficient for ten calibrations.