Amino acid F reagent
Lieferant: Hach
2386442
HACH2386442EA 30.9 EUR
HACH2386442
Amino acid F reagent
Umwelt-Prüfkits
Reagent for the determination of ultra low range silica by the heteropoly blue method. Hach method range: 3 to 1000 µg/L SiO₂.