Ampoule snapper, AccuVac®
Lieferant: Hach
2405200
HACH2405200EA 29.4 EUR
HACH2405200
Ampoule snapper, AccuVac®
Ampullenöffner
This snapper is used for opening ampoules immersed in samples.
  • Avoids sample contact
  • Used with colorimeters and spectrophotometers