Nitrogen-nitrate standard solution
Lieferant: Hach
2415132
1426010
194749
204649
HACH2415132EA
17.7
EUR
HACH2415132
705-2010
705-2025
HACH204649
Nitrogen-nitrate standard solution
Standards
Umweltanalyse-Standards
Standard solutions for accuracy checks in nitrate nitrogen determinations and also for meter calibrations/standardisation.