Reagent set, glycol test kit
Lieferant: Hach
2446900
HACH2446900EA
594
EUR
HACH2446900
Reagent set, glycol test kit
Umwelt-Prüfkits
Used for determination of glycol by the EG-1 glycol test kit.
Lieferumfang: Reagent set includes: glycol test reagent 1, glycol test reagent 2, glycol test reagent 3 and ethylene glycol - 0.05% solution.