Test kits, digital titration method
Lieferant: Hach
2448600 2064100 2063500 1438801 1439301 1496101 83699 2063700 2063600 2288000 2081701 2081801 1449501 1439701 1439601 1440301 1440201 2267601 2471100 2445300 2272700 1440101 2580600 1439101 2272600 2447500 2448100 2272300 2292301 2393701 2686901 2272200 2448000 92101 2447200 2449300 2447000 2062501 2409301 1439001
HACH2448600EA 149 EUR
HACH2448600 HACH2064100 HACH2063500 HACH1438801 705-0066 705-0074 705-0240 705-0082 HACH2063600 HACH2288000 HACH2081701 HACH2081801 HACH1449501 HACH1439701 HACH1439601 HACH1440301 HACH1440201 705-0115 HACH2471100 HACH2445300 HACH2272700 HACH1440101 HACH2580600 HACH1439101 HACH2272600 HACH2447500 HACH2448100 HACH2272300 HACH2292301 HACH2393701 HACH2686901 HACH2272200 HACH2448000 HACH92101 HACH2447200 HACH2449300 HACH2447000 HACH2062501 HACH2409301 HACH1439001
Test kits, digital titration method
Umwelt-Prüfkits
Digital titration provides a more precise and accurate method than the drop test method.
  • Reagents are pre-prepared
  • Exact comparison of colour
  • Rugged carrying case for analysis in the field
Bitte beachten Please note some of the reagents supplied in the kits are hazardous, and appropriate precautions should be taken for use and disposal. Please contact Avantor for details.