Glass microfibre filters without binder
Lieferant: Hach
253000
HACH253000EA 73.2 EUR
HACH253000
Glass microfibre filters without binder
Filter Glasfaserfilter
These binder free TSS glass microfibre filters are made of 100% borosilicate glass and are designed for total suspended solids analysis of water and wastewater. They are recommended for use in Standard Methods 2540 C and 2540 D.
  • Optimised surface characteristics eliminates pin-holing and fiber-shedding to ensure consistent, accurate results
  • High flow rate, fine porosity