Hydrogen sulfide test kit, HS-C
Lieferant: Hach
2537800
HACH2537800EA 0 EUR
HACH2537800
Hydrogen sulfide test kit, HS-C
Umwelt-Prüfkits
The kit is used to determine sulphide pollution of sewage water, waste water and other sources resulting from decomposition of organic matter. In the HS-C kit, sulphide discolours a detection paper. The results can be obtained by comparison of the colour to a printed chart.
  • Reliable, cost effective colour comparison method
Lieferumfang: Includes reaction vial, effervescent tablets, test papers, comparator chart and instructions, in a cardboard box.