Ammonia electrode storage solution
Lieferant: Hach
2541249
HACH2541249EA 60.8 EUR
HACH2541249
Ammonia electrode storage solution
Elektrolyte Elektrodenaufbewahrungslösungen
For storage of ammonia ion selective electrodes.