Bromthymolblau

Lieferant: Hach

25532
HACH25532EA 21.8 EUR
HACH25532
Bromthymolblau
Bromthymolblau
Formel: C₂₇H₂₈Br₂O₅S
Schmelzpunkt: 204…202 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00005872
CAS-Nummer: 76-59-5
EINECS: 200-971-2
Merck Index: 13,01430

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR