Thymolblau -Lösung in 2-Propanol/ Wasser (2:3) Indikator

Lieferant: Hach
Gefahr

Synonyme: 4-[3-(4-Hydroxy-2-methyl-5-propan-2-ylphenyl)-1,1-dioxobenzo[c]oxathiol-3-yl]-5-methyl-2-propan-2-ylphenol, Thymolsulfonphthalein

25732
HACH25732EA 27.9 EUR
HACH25732
Thymolblau -Lösung in 2-Propanol/ Wasser (2:3) Indikator
Thymolblau

Solution for colorimetric pH determination of pH range: 7,8 - 10,0.

Formel: C₂₇H₃₀O₅S
Molecular Weight: 466.6 g/mol
Schmelzpunkt: 221
MDL: MFCD00005869
CAS-Nummer: 76-61-9
UN: 1219
ADR: 3 II
Merck Index: 12,09541

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR