pH indicator strips
Lieferant: Hach
2601300
HACH2601300EA 51 EUR
HACH2601300
pH indicator strips
Testpapiere pH-Indikatorstreifen
pH strips provides a simple, fast and economical method of pH measurement. Can obtain accurate results in weakly buffered, turbid and lightly coloured samples. It is easy to use and disposable.