Natriumthiosulfat -Lösung 2.12 N

Lieferant: Hach

Synonyme: di-Natriumthiosulfat, Antichlor, Fixiersalz, Natriumhyposulfit

2686832
HACH2686832EA 51.8 EUR
HACH2686832
Natriumthiosulfat -Lösung 2.12 N
Natriumthiosulfat

For determination of high range hypochlorite (bleach) concentrations by drop-count titration.

Formel: Na₂S₂O₃
Molecular Weight: 158.11 g/mol
Siedepunkt: 100
Schmelzpunkt: 48
MDL: MFCD00003499
CAS-Nummer: 7772-98-7
Merck Index: 12,08844

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR