Turbidity standard, ULR verification kit, Stablcal®
Lieferant: Hach
2714600
HACH2714600EA
297
EUR
HACH2714600
Turbidity standard, ULR verification kit, Stablcal®
Standards
Trübungsstandards
Stablcal® verification kit containing 0.10 NTU, 0.30 NTU, 0.50 NTU, 1.0 NTU standards (100 ml each), sililcone oil, oiling cloth and instructions is used for verification purpose only.
- Precise, pre-mixed formazin solutions
- Enhanced stability saves time and labor
- Confidence in results