THM reagent 3
Lieferant: Hach
2754142
HACH2754142EA
170
EUR
HACH2754142
THM reagent 3
Umwelt-Prüfkits
Screening tool to optimise treatment processes, to locate areas of high THM levels in distribution systems and to determine levels of trihalomethanes in drinking water samples.
- For determination of Total Trihalomethanes by the THM Plus™ Method
- Hach Method 10132
Bitte beachten Corrosive