Oxygen demand, quality control standard
Lieferant: Hach
2833510
HACH2833510EA 89.1 EUR
HACH2833510
Oxygen demand, quality control standard
Standards Umweltanalyse-Standards
These mixed standards are ready to use, no dilutions are required. They contain BOD, COD and TOC.
Bestellinformation: Pack of 16×10 ml ampoules.