Colour disc test kit, chlorine dioxide
Lieferant: Hach
2842800
HACH2842800EA 173 EUR
HACH2842800
Colour disc test kit, chlorine dioxide
Umwelt-Prüfkits
The chlorine dioxide test kit uses the reliable and EPA-accepted DPD method to monitor ClO2 concentration, while eliminating chlorine interference through the addition of glycine.
  • Simple, cost-effective method for maintaining optimum chlorine dioxide levels in your plant
  • USEPA-accepted for waste water and potable water
With available range of 0 to 2 mg/l and resolution of 0,1 mg/l, operators can accurately monitor around the typical 0,8 mg/l regulated limit.
Lieferumfang: Kit includes DPD powder pillows, glycine reagent, plastic viewing tube, colour comparator box, colour discs, instruction sheets, and carrying case.