Filling Solution, Reference, 1 M KNO₃
Lieferant: Hach
28564026
HACH28564026EA 23.2 EUR
HACH28564026
Filling Solution, Reference, 1 M KNO₃
Elektrolyte Elektrodenfülllösungen
Filling solution for electrode reference element, potassium nitrate (KNO₃), 1 M solution.