BOD seed inoculum, POLYSEED
Lieferant: Hach
2918700
HACH2918700EA 332 EUR
HACH2918700
BOD seed inoculum, POLYSEED
Mikrobiologische Testsysteme Tests zur Überprüfung von Mirkoorganismen
BOD seed inoculum for use in biochemical oxygen demand (BOD5) test procedures. A blend of specialised microbial cultures in an easy-to-use capsule. Designed to provide a uniform standard for the degradation of both industrial and municipal waste in BOD5 analyses.
Bestellinformation: Package of 50 capsules. International Laboratory Supply (InterLab), LTD.