Trübungsstandard-Kalibrierkit für 2100Q, Stablcal®
Lieferant: Hach
2659405
2659410
2659400
2961701
2971205
2971210
2971200
707-0011EA
408
EUR
707-0011
HACH2659410
HACH2659400
HACH2961701
705-0925
HACH2971210
HACH2971200
Trübungsstandard-Kalibrierkit für 2100Q, Stablcal®
Standards
Trübungsstandards
Stablcal® calibration standards kit, for 2100 series portable turbidimeters. USEPA accepted for calibration purposes. Stablcal® stabilised formazin turbidity standards are true formazin dilutions developed for use in any turbidimeter.
- Precise, pre-mixed formazin solutions
- Based on formazin primary standards
- Enhanced stability saves time and labour
- Confidence in results