Trübungsstandard-Kalibrierkit für 2100Q, Stablcal®
Lieferant: Hach
2659405 2659410 2659400 2961701 2971205 2971210 2971200
707-0011EA 408 EUR
707-0011 HACH2659410 HACH2659400 HACH2961701 705-0925 HACH2971210 HACH2971200
Trübungsstandard-Kalibrierkit für 2100Q, Stablcal®
Standards Trübungsstandards
Stablcal® calibration standards kit, for 2100 series portable turbidimeters. USEPA accepted for calibration purposes. Stablcal® stabilised formazin turbidity standards are true formazin dilutions developed for use in any turbidimeter.
  • Precise, pre-mixed formazin solutions
  • Based on formazin primary standards
  • Enhanced stability saves time and labour
  • Confidence in results