N,N-Diethyl-p-phenylenediamin oxalat analytisches Reagens

Lieferant: Hach

Synonyme: Bis(N,N-diethylbenzene-p-diamine) oxalate, DPD oxalate

608424
HACH608424EA 153 EUR
HACH608424
N,N-Diethyl-p-phenylenediamin oxalat analytisches Reagens
N,N-Diethyl-p-phenylenediamine oxalate
Formel: C₁₀H₁₆N₂·0.5 C₂H₂O₄
Molecular Weight: 209,27 g/mol
MDL: MFCD00070242
CAS-Nummer: 62637-92-7

Bestellen

Spezifikation Testergebnisse

Assay 65 - 95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR