Hydroxylammoniumchlorid 10% in wässriger Lösung

Lieferant: Hach
Achtung

Synonyme: Hydroxylamin-Hydrochlorid, Chlorhydrathydroxylamin, Hydroxylamin Hydrochlorid, Hydroxylamin HCl

81842
HACH81842EA 23.3 EUR
HACH81842
Hydroxylammoniumchlorid 10% in wässriger Lösung
Hydroxylammoniumchlorid
Formel: NH₂OH·HCl
Molecular Weight: 69,49 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00051089
CAS-Nummer: 5470-11-1
EINECS: 226-798-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR