Combined pH electrode, red rod
Lieferant: Hach
945-252
HACH945-252EA 472 EUR
HACH945-252
Combined pH electrode, red rod
Elektroden pH-Elektroden
The Radiometer GK2401C combination pH electrode has a glass body, Red-Rod reference system, refillable electrolyte (saturated KCl) and Radiometer Type 7 connector for compatibility with Radiometer MeterLab legacy meters.
  • Red-Rod electrodes provide a stable signal fast by protecting the reference elements from light interference
  • The Red-Rod saturated KCl salt-bridge also aids in creating a stable liquid junction potential
  • Red-Rod system creates a trouble-free liquid junction: No silver ions and therefore less risk of clogging when measuring in challenging sample matrices
  • Red-Rod reference system enables use over a wider range of temperatures than for most pH electrodes