Pipetten-Set
Lieferant: Hach
LZP320
HACHLZP320EA 781 EUR
HACHLZP320
Pipetten-Set
Pipetten Mikropipetten
These variable volume single-channel, air-displacement pipettes feature outstanding accuracy with adjustments in 50 µl increments.
  • Robust design
  • Comfortable ergonomics
  • Slim-line tip ejection
  • Single-handed operation
Ideal for use with HACH TNTplus reagents.
Bestellinformation: Supplied with calibration report and maintenance tool.