ORP electrode, sensION+ 5057
Lieferant: Hach
LZW5057970002
HACHLZW5057970002EA 381 EUR
HACHLZW5057970002
ORP electrode, sensION+ 5057
Elektroden Redox-Elektroden
sensION+ 5057 is a combination platinum ORP electrode with a glass body and refillable liquid-electrolyte reference. It is dedicated for use with Hach sensION+ labpratory pH (mV) meters.
  • Fixed 1 meter cable and BNC connector
  • Universal design for a variety of applications
  • Encapsulated cartridge reference system ensures premium stability and long lifespan
  • Innovative reference system utilises a “cartridge” of Ag/AgCl crystals contacting a silver wire inside a glass tube
The 5057 is ideal for oxidation-reduction potential (ORP) measurements in general aqueous applications.
Lieferumfang: Supplied with 3,0 M KCl electrolyte refill solution (30 ml) and basic user manual.