Multi-parameter electrode, sensION+ 5059
Lieferant: Hach
LZW5059970002
HACHLZW5059970002EA 495 EUR
HACHLZW5059970002
Multi-parameter electrode, sensION+ 5059
Elektroden Multiparameter-Elektroden
sensION+ 5059 is a combination multi-parameter electrode for determination of pH, conductivity and temperature. It has a plastic body, non-refillable gel reference electrolyte and built-in temperature sensor.
  • Determine multiple parameters in the field with one standard sized electrode
  • Low maintenance design for a variety of applications
  • Ultra portable but protected against harsh field conditions
  • Heavy-duty electrode handle design optimised for field calibration and storage
  • Glass (pH); platinum (conductivity); and Pt1000 (temperature) sensors
Dedicated for use with Hach sensION+ MM150 portable meters. Ideal for pH, conductivity and temperature measurements in general aqueous applications.
Lieferumfang: Supplied with basic user manual.