Ion selective electrode, half-cell, chloride, sensION+ 9652
Lieferant: Hach
LZW9652970002
HACHLZW9652970002EA 1085 EUR
HACHLZW9652970002
Ion selective electrode, half-cell, chloride, sensION+ 9652
Elektroden ISE-Elektroden
sensION+ 9652 is a half-cell (reference not integrated) chloride ion selective electrode with an epoxy body and non-replaceable crystalline membrane selective to chloride ions in solution.
  • Utilises a unique solid-state sensor technology that eliminates costly membrane replacements
  • Requires virtually no maintenance: Solid-state sensor design allows for dry storage of the ISE without a shelf life or membrane replacement
  • Provides fast, stable, and accurate response in a variety of sample types
The 9652 has a fixed 1 meter cable and BNC connector for laboratory use with the Hach sensION+ MM374 Multimeter. The 9652 is ideal for measuring chloride concentrations in wastewater, drinking water and general water quality applications.
Lieferumfang: Supplied with basic user manual.