Ion selective electrode, half-cell, fluoride, sensION+ 9655
Lieferant: Hach
LZW9655970002
HACHLZW9655970002EA 1053 EUR
HACHLZW9655970002
Ion selective electrode, half-cell, fluoride, sensION+ 9655
Elektroden ISE-Elektroden
sensION+ 9655 is a half-cell (reference not integrated) fluoride ion selective electrode with an epoxy body and non-replaceable crystalline membrane selective to fluoride ions in solution.
  • Requires virtually no maintenance: Solid-state sensor design allows for dry storage of the ISE without a shelf life or membrane replacement
  • Provides fast, stable, and accurate response in a variety of sample types
  • Recommended for use with the sensION+ 5044 reference electrode
The 9655 has a fixed 1 meter cable and BNC connector for laboratory use with the Hach sensION+ MM374 Multimeter. The electrode is ideal for measuring fluoride concentrations in drinking water, wastewater and general water quality applications.
Lieferumfang: Supplied with basic user manual.