Automatic vacuum aspiration systems, multifunctional, AA 02
Lieferant: HAEP LABOR
981323101
HAEP981323101EA 2310 EUR
HAEP981323101
Automatic vacuum aspiration systems, multifunctional, AA 02
Pumpen Vakuum-Absaugsysteme
Automatic suction system with suction capacity for connection to laboratory vacuum system.
Bestellinformation: System consist of vacuum pump (100 mbar) with integral bottle holder, plastic safety bottle (2 L), silicone tubing (3 m), disposable filter to protect pump and environment.
Lieferumfang: Supplied with AH01 handle.