Automatic vacuum aspiration systems, AA 24
Lieferant: HAEP LABOR
981323201
HAEP981323201EA 2620 EUR
HAEP981323201
Automatic vacuum aspiration systems, AA 24
Pumpen Vakuum-Absaugsysteme
Automatic safety vacuum system for extraction.
  • Standard for two users
  • Extremely robust metal housing
  • Suction power of 17 L air/min, absolute vacuum: 100 mbar
  • Polypropylene bottle includes quick connect couplings, filters, ventilation insert and 3 meter silicone hoses