Refrigerated circulating bath, VersaCool™
Lieferant: THERMO ELECTRON LED
231131300
HAKA231131300EA 12100 EUR
HAKA231131300
Refrigerated circulating bath, VersaCool™
Umwälzthermostate Geschlossene Umwälzsysteme Kälteumwälzthermostate
The VersaCool™ Refrigerated circulating bath features a new headless design, suitable for use as a recirculating refrigerated bath or heated circulator to circulate fluid from the bath to specific applications.
  • More space to fit samples
  • Automatically detects appropriate voltage and works on all voltages worldwide
  • Lower power consumption and energy costs
  • Tool-less, drip-less rack
  • Left-handed hinge, right-handed hinge, or unhinged lid
  • Intuitive, 5,7" touchscreen colour display for easier and more efficient setup
  • Two year warranty
  • Ethernet RS232
It can easily be used on or under a bench, fume hood, industrial and even mobile installations. Advancements in remote monitoring capabilities, energy efficiency, global voltage input, and other tools and features give you control and optimal precision.
This water bath continually operates in an energy efficient mode abnd has variable speed control in pumps, fans and compressors.
The rack accommodates a wide range of beaker and test tube sizes. The dripless lid and optional gable cover reduce moisture and directs condensate back into bath.
The communication and control features ensures that samples are properly maintained.