Circulating water baths, Precision™
Lieferant: THERMO ELECTRON LED
TSCIR35
HAKATSCIR35EA 2850 EUR
HAKATSCIR35
Circulating water baths, Precision™
Bäder Schüttelwasserbäder
These circulating water baths are ideal when temperature uniformity and control are particularly critical, such as when working with enzymes and in serological applications.
  • Enhanced temperature uniformity with perimeter-directed water flow
  • Easily clean and maintain bath with coil-free internal design
  • Easily operate and monitor with icon-based graphical display
  • Prevent bath overheating and damage with low-fluid protection
  • Monochrome LCD display
Lieferumfang: Supplied with stainless steel gable cover and rubber duck.