General purpose waterbaths, Precision™, with metal balls
Lieferant: THERMO ELECTRON LED
TSGP2S
HAKATSGP2SEA 830 EUR
HAKATSGP2S
General purpose waterbaths, Precision™, with metal balls
Bäder Wasserbäder, ohne Umwälzung
The Precision™ range of waterbaths is ideal for a wide range of applications, offers the choice of working in 'water' or 'bead' mode. Designed to work at temperatures up to 100 °C, these baths have capacities from 2 up to 28 litres**. Uniformity of temperature is the same at 37 °C, for a 5 L bath, in 'water' or 'ball' mode: ± 0,2 °C.
  • Over-temperature protection prevents thermal drift, while automatic start/ stop timers help maximize productivity
  • Excellent chemical and corrosion resistance thanks to epoxy powder coated exterior
  • Tank is easy to clean thanks to its seamless stainless steel inner surface
  • Simplified use with quick-disconnect vent on 10, 20 and 28 litre models as well as double bowl models
Lieferumfang: Supplied with a transparent polycarbonate hinged lid, as well as a diffuser tray and a drain hose / to be used for 'water' mode, and one or more bead bags (depending on models) / to be used for the mode 'balls'.
Bitte beachten * RT = Ambient
** Filling of baths: at least 3 cm high of logs or water and at most 4 cm below the rim of the bath tub.