pH Cleaning Solution Set
Lieferant: HAMILTON BONADUZ
A two-part cleaning solution for all glass membrane pH sensors. pH Cleaning solution A contains sodium hydroxide (<3%), pH cleaning solution B contains hydrochloric acid (4%).
Also includes storage solution.
Lieferumfang: Includes 2 × 500 ml A+B cleaning solutions + 1 × 500 ml storage solution.