Dissecting scissors
Lieferant: HAMMACHER KARL
HSB300-11 HSB301-11
HAMMHSB300-11EA 284 EUR
HAMMHSB300-11 HAMMHSB301-11
Dissecting scissors
Scheren
These dissecting scissors made of stainless steel feature tungsten carbide inserts.