Bandage scissors, Lister type
Lieferant: HAMMACHER KARL
HSB587-20 HSB585-18 HSB583-14
233-1153EA 95.1 EUR
233-1153 HAMMHSB585-18 HAMMHSB583-14
Bandage scissors, Lister type
Scheren
These stainless steel scissors are ideal for sizing dressings and removing bandages.
The jaws of the Lister scissors are angled with the lower blade being slightly longer. The tip of the lower blade features a flattened blunt nodule that is intended to slide between bandages and skin without cutting the skin.