Crown stopwatch, pulse/breathing rate
Lieferant: HANHART
112.7001-00
HANH112.7001-00EA 370 EUR
HANH112.7001-00
Crown stopwatch, pulse/breathing rate
Timer Stoppuhren, analog
Precision mechanics with mineral glass and metal crown. Specially for measuring pulse and breathing rate. Based on 15 pulse beats or 5 breaths.
  • Timing capacity: 30 minutes (1/10 seconds)
  • Pin lever, 7 jewels
  • Glass parts are resistant to scratching and acid
  • Metal crown is extremely resistant to solvents and thinners
Ideal for use in the chemical industry, paint workshops or photographic laboratories.
Weight: 120 g