Crown stopwatch, Robust 100
Lieferant: HANHART
115.0104-0S
HANH115.0104-0SEA 579 EUR
HANH115.0104-0S
Crown stopwatch, Robust 100
Timer Stoppuhren, analog
Precision stopwatch with mineral glass and metal crown.
  • Crown stopper with pin lever and 7 jewels
  • Glass parts are resistant to scratching and acid
  • Metal crown is extremely resistant to solvents and thinners
  • Especially suitable for use in the chemical industry, paint workshops, or photographic laboratories
Stopwatch was tested in a 30 degree, crown up position.