Electrode storage solution
Lieferant: Hanna
HI70300L
HANNHI70300LEA 20.7 EUR
HANNHI70300L
Electrode storage solution
Elektrolyte Elektrodenaufbewahrungslösungen
HI 70300L is a storage solution prepared with reagent grade chemicals which can be used to ensure maximum performance of pH and ORP electrodes.
  • Keeps the glass membrane of the pH electrode hydrated
  • Manufactured specifically for pH and ORP electrodes
  • Air tight bottle with tamper-proof seal ensures quality and freshness of solution
  • Clearly marked expiration date and lot number
The storage solution is specifically formulated to minimise microbial growth and to prevent any effects of diffusion/osmotic from storing a probe in a solution with the highly concentrated inner reference electrolyte.